Learn More
PLA2G3 Mouse anti-Human, Clone: OTI4F2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811652

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
PLA2G3 belongs to the family of secreted phospholipase A2 (sPLA2; EC 3.1.1.4) proteins. These Ca(2+)-dependent lipolytic enzymes have a conserved Ca(2+)-binding loop and a his-asp dyad in the catalytic site.Specifications
PLA2G3 | |
Monoclonal | |
Unconjugated | |
PLA2G3 | |
GIII-SPLA2, SPLA2III, sPLA2-III | |
Mouse | |
Affinity Chromatography | |
RUO | |
50487 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4F2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NZ20 | |
PLA2G3 | |
Human recombit protein fragment corresponding to amino acids 409-509 of human PLA2G3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.