Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLA2G3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582742
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582742 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582742 Supplier Invitrogen™ Supplier No. PA582742
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPPEVTNMLW ELLGTTCFKL APPLDCVEGK NCSRDPRAIR VSARHLRRLQ QRRHQLQDKG TDERQPWPSE PLRGPMSFYN QCLQLTQAAR RPDRQQKSWS Q Highest antigen sequence indentity to the following orthologs: Rat - 63%, Mouse - 60%.

PLA2G3 belongs to the family of secreted phospholipase A2 (sPLA2; EC 3.1.1.4) proteins. These Ca(2+)-dependent lipolytic enzymes have a conserved Ca(2+)-binding loop and a his-asp dyad in the catalytic site.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2G3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLA2G3
Gene Accession No. Q9NZ20
Gene Alias giii; GIII sPLA2; GIII-SPLA2; Group 3 secretory phospholipase A2; group III secreted phospholipase A2; Group III secretory phospholipase A2; Phosphatidylcholine 2-acylhydrolase 3; phosphatidylcholine 2-acylhydrolase GIII; phospholipase A2 group III; phospholipase A2, group III; PLA2G3; SPLA2III; sPLA2-III
Gene Symbols PLA2G3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLA2G3. Recombinant protein control fragment (Product #RP-92349).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50487
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.