Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLA2G4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550642
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550642 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550642 Supplier Invitrogen™ Supplier No. PA550642
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total PLA2G4B protein.

PLA2G4B transcribes naturally-occurring mRNAs that are co-transcribed products of the neighboring JMJD7 and PLA2G4B genes. Incompletely processed read-through transcripts from these two loci are abundantly expressed in most tissues, but the function of the predicted protein product has not yet been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2G4B
Applications Western Blot
Classification Polyclonal
Concentration 2.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene PLA2G4B
Gene Accession No. P0C869, P0C871
Gene Alias A030011C02Rik; cPLA2-beta; cytosolic phospholipase A2 beta; Gm1013; HsT16992; JMJD7-PLA2G4B; JMJD7-PLA2G4B protein; JMJD7-PLA2G4B readthrough; jumonji domain containing 7-phospholipase A2, group IVB (cytosolic) read-through; phospholipase A2 group IVB; phospholipase A2, group IVB (cytosolic); Pla2g4b
Gene Symbols PLA2G4B
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from internal residues of human phospholipase A2, group IVB (cytosolic).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100137049, 211429
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.