Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLA2G4B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103775
Description
Antibody detects endogenous levels of total PLA2G4B.
PLA2G4B transcribes naturally-occurring mRNAs that are co-transcribed products of the neighboring JMJD7 and PLA2G4B genes. Incompletely processed read-through transcripts from these two loci are abundantly expressed in most tissues, but the function of the predicted protein product has not yet been determined.
Specifications
PLA2G4B | |
Polyclonal | |
Unconjugated | |
PLA2G4B | |
A030011C02Rik; cPLA2-beta; cytosolic phospholipase A2 beta; Gm1013; HsT16992; JMJD7-PLA2G4B; JMJD7-PLA2G4B protein; JMJD7-PLA2G4B readthrough; jumonji domain containing 7-phospholipase A2, group IVB (cytosolic) read-through; phospholipase A2 group IVB; phospholipase A2, group IVB (cytosolic); Pla2g4b | |
Rabbit | |
Affinity chromatography | |
RUO | |
100137049, 211429 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P0C869, P0C871 | |
PLA2G4B | |
A synthesized peptide derived from human PLA2G4B(Accession P0C869), corresponding to amino acid residues L644-A694. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction