Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLA2G4C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560231
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560231 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560231 Supplier Invitrogen™ Supplier No. PA560231
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EAELDLWSKA PASCYILKGE TGPVVMHFPL FNIDACGGDI EAWSDTYDTF KLADTYTLDV VVLLLALAKK NVRENKKKIL RELMNVAGLY YPKD Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 53%.

This gene encodes a protein which is a member of the phospholipase A2 enzyme family which hydrolyzes glycerophospholipids to produce free fatty acids and lysophospholipids, both of which serve as precursors in the production of signaling molecules. The encoded protein has been shown to be a calcium-independent and membrane bound enzyme. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Apr 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2G4C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLA2G4C
Gene Accession No. Q9UP65
Gene Alias cPLA2-gamma; Cytosolic phospholipase A2 gamma; DKFZp586C0423; FLJ42247; FLJ44164; phospholipase A2 group IVC; phospholipase A2, group IVC (cytosolic, calcium-independent); PLA2G4C
Gene Symbols PLA2G4C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLA2G4C. Recombinant protein control fragment (Product #RP-98985).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8605
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.