Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLA2G4C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596790
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596790 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596790 Supplier Invitrogen™ Supplier No. PA596790
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGSSEVSIIP GLQKEEKAAV ERRRLHVLKA LKKLRIEADE APVVAVLGSG GGLRAHIACL GVLSEMKEQG LLDAVTYLAG VSGSTWAISS LYTNDGDMEA LEADLKHRFT RQEWDLAKSL QKTIQAARSE NYSLTDFWAY MVISKQTREL PESHLSNMKK PVEEGTLPYP IFAAIDNDLQ PSWQEARAP; Positive Samples: THP-1; Cellular Location: Lipid-anchor, Membrane.

This gene encodes a protein which is a member of the phospholipase A2 enzyme family which hydrolyzes glycerophospholipids to produce free fatty acids and lysophospholipids, both of which serve as precursors in the production of signaling molecules. The encoded protein has been shown to be a calcium-independent and membrane bound enzyme. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Apr 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2G4C
Applications ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 2.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PLA2G4C
Gene Accession No. Q9UP65
Gene Alias cPLA2-gamma; Cytosolic phospholipase A2 gamma; DKFZp586C0423; FLJ42247; FLJ44164; phospholipase A2 group IVC; phospholipase A2, group IVC (cytosolic, calcium-independent); PLA2G4C
Gene Symbols PLA2G4C
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-189 of human PLA2G4C (NP_0036972).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8605
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.