Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLA2G4D Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA551226

Catalog No. PA551226


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total PLA2G4D / cPLA2-delta protein.

The phospholipase A2 enzyme family, including PLA2G4D, catalyze the hydrolysis of glycerophospholipids at the sn-2 position and then liberate free fatty acids and lysophospholipids.[supplied by OMIM, Jun 2009].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PLA2G4D
Polyclonal
Unconjugated
PLA2G4D
2610311B01Rik; cPLA2delta; cPLA2-delta; cytosolic phospholipase A2 delta; phospholipase A2 delta, cytosolic; phospholipase A2 group IVD; phospholipase A2, group IVD; phospholipase A2, group IVD (cytosolic); Pla2delta; PLA2G4D
Rabbit
Antigen affinity chromatography
RUO
283748
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.8 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q86XP0
PLA2G4D
Fusion protein of human PLA2G4D.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.