Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLA2G4D Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572287
Description
PLA2G4D Polyclonal Antibody for Western Blot
The phospholipase A2 enzyme family, including PLA2G4D, catalyze the hydrolysis of glycerophospholipids at the sn-2 position and then liberate free fatty acids and lysophospholipids.[supplied by OMIM, Jun 2009].
Specifications
PLA2G4D | |
Polyclonal | |
Unconjugated | |
PLA2G4D | |
2610311B01Rik; cPLA2delta; cPLA2-delta; cytosolic phospholipase A2 delta; phospholipase A2 delta, cytosolic; phospholipase A2 group IVD; phospholipase A2, group IVD; phospholipase A2, group IVD (cytosolic); Pla2delta; PLA2G4D | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
283748 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q86XP0 | |
PLA2G4D | |
KLH conjugated synthetic peptide between 754-786 amino acids from human PLA2G4D. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction