Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLA2G4D Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145068
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145068 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145068 Supplier Invitrogen™ Supplier No. PA5145068
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLKGSYEDTQ TSFLGTASAF RFHYMAALET ELSGRLRSSR SNGWNGDNSA GYLTVPLRPL TIGKEVTMDV PAPNAPGV Antigen sequence identity: Mouse 55%; Rat 55%.

The phospholipase A2 enzyme family, including PLA2G4D, catalyze the hydrolysis of glycerophospholipids at the sn-2 position and then liberate free fatty acids and lysophospholipids.[supplied by OMIM, Jun 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2G4D
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLA2G4D
Gene Accession No. Q86XP0
Gene Alias 2610311B01Rik; cPLA2delta; cPLA2-delta; cytosolic phospholipase A2 delta; phospholipase A2 delta, cytosolic; phospholipase A2 group IVD; phospholipase A2, group IVD; phospholipase A2, group IVD (cytosolic); Pla2delta; PLA2G4D
Gene Symbols PLA2G4D
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 188-265. Recombinant protein control fragment (Product #RP-109863).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283748
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.