Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLA2G4E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549553
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549553 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549553 Supplier Thermo Scientific Supplier No. PA549553
Add to Cart
Add to Cart

PLA2G4E Polyclonal Antibody

PLA2G4E Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2G4E
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene Pla2g4e
Gene Accession No. Q3MJ16
Gene Alias 2310026J01Rik; C230096D22; cPLA2-epsilon; cytosolic phospholipase A2 epsilon; EC 3.1.1.4; Phospholipase A2 group IVE; phospholipase A2, group 4E; phospholipase A2, group IVE; Pla2epsilon; Pla2g4e; RGD1310595
Gene Symbols Pla2g4e
Host Species Rabbit
Immunogen synthetic peptide corresponding to 428-456aa of human PA24E.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123745
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.