Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLA2G6 Antibody (OTI10G4), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273456H
Description
PLA2G6 Monoclonal specifically detects PLA2G6 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PLA2G6 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
PLA2G6 | |
Recombinant protein fragment corresponding to amino acids 490-752 of human PLA2G6 (NP_001004426) produced in E.coli. | |
0.1 mL | |
8398.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI10G4 | |
Western Blot | |
calcium-independent phospholipase A2, EC 3.1.1.4, Group VI phospholipase A2, IPLA2, iPLA285 kDa calcium-independent phospholipase A2, IPLA2-VIA, NBIA2A, NBIA2B, PARK14GVI, patatin-like phospholipase domain containing 9, phospholipase A2, group VI (cytosolic, calcium-independent), PNPLA9 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction