Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLA2G6 Polyclonal Antibody

PLA2G6 Polyclonal Antibody

Supplier:  Thermo Scientific PA550637

Catalog No. PA550637


Only null left
Add to Cart

Description

Description

PLA2G6 Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

PLA2G6
Polyclonal
Unconjugated
PLA2G6
85 kDa calcium-independent phospholipase A2; 85/88 kDa calcium-independent phospholipase A2; BB112799; CaI-PLA2; Group VI phospholipase A2; GVI; GVI PLA2; INAD1; intracellular membrane-associated calcium-independent phospholipase A2 beta; iPLA(2)beta; iPLA2; iPLA2beta; iPLA2-beta; IPLA2-VIA; NBIA2; NBIA2A; NBIA2B; neurodegeneration with brain iron accumulation 2; PARK14; patatin-like phospholipase domain-containing protein 9; phospholipase A2 group VI; phospholipase A2, group VI; phospholipase A2, group VI (cytosolic, calcium-independent); PLA2; PLA2G6; PLPLA9; PNPLA9
Rabbit
Antigen Affinity Chromatography
RUO
360426, 53357, 8398
-20°C
Liquid
Immunohistochemistry (Paraffin)
1.3 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
O60733, P97570, P97819
PLA2G6
Synthetic peptide corresponding to a region derived from internal residues of human phospholipase A2, group VI (cytosolic, calcium-independent).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.