Learn More
Invitrogen™ PLA2G6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551525
Description
Immunogen sequence: QVLHTEVLQH LTDLIRNHPS WSVAHLAVEL GIRECFHHSR IISCANCAEN EEGCTPLHLA CRKGDGEILV ELVQYCHTQM DVTDYKGETV FHYAVQGDNS QVLQLLGRNA VAGLNQVNNQ GLTPLHLACQ LGKQEMVRVL LLCNARCNIM G Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
The protein encoded by this gene is an A2 phospholipase, a class of enzyme that catalyzes the release of fatty acids from phospholipids. The encoded protein may play a role in phospholipid remodelling, arachidonic acid release, leukotriene and prostaglandin synthesis, fas-mediated apoptosis, and transmembrane ion flux in glucose-stimulated B-cells. Several transcript variants encoding multiple isoforms have been described, but the full-length nature of only two of them have been determined to date.
Specifications
| PLA2G6 | |
| Polyclonal | |
| Unconjugated | |
| PLA2G6 | |
| 85 kDa calcium-independent phospholipase A2; 85/88 kDa calcium-independent phospholipase A2; BB112799; CaI-PLA2; Group VI phospholipase A2; GVI; GVI PLA2; INAD1; intracellular membrane-associated calcium-independent phospholipase A2 beta; iPLA(2)beta; iPLA2; iPLA2beta; iPLA2-beta; IPLA2-VIA; NBIA2; NBIA2A; NBIA2B; neurodegeneration with brain iron accumulation 2; PARK14; patatin-like phospholipase domain-containing protein 9; phospholipase A2 group VI; phospholipase A2, group VI; phospholipase A2, group VI (cytosolic, calcium-independent); PLA2; PLA2G6; PLPLA9; PNPLA9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8398 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O60733 | |
| PLA2G6 | |
| Recombinant protein corresponding to Human PLA2G6. Recombinant protein control fragment (Product #RP-91351). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.