Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLA2R1 Antibody (12-6-5), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250248AF350
Description
PLA2R1 Monoclonal specifically detects PLA2R1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
PLA2R1 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
PLA2R1 | |
Human PLA2R extracellular sequence N-C3 (containing domains N-terminal Cysteine rich-FibII-CTLD1-CTLD2-CTLD3). Genbank acc no. U17033; amino acids 20-663 Recombinant protein expressed in HEK 293-EBNA-1 cells | |
Primary | |
Human | |
IgG2b |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
38692 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
180 kDa secretory phospholipase A2 receptor, CLEC13CC-type lectin domain family 13 member C, phospholipase A2 receptor 1, 180kDa, PLA2G1R, PLA2IR, PLA2-RM-type receptor, PLA2Rphospholipase A2 receptor 1, 180kD, secretory phospholipase A2 receptor | |
Mouse | |
0.1 mL | |
22925 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction