Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLA2R1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552966
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552966 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552966 Supplier Invitrogen™ Supplier No. PA552966
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEKTWHEALR SCQADNSALI DITSLAEVEF LVTLLGDENA SETWIGLSSN KIPVSFEWSN DSSVIFTNWH TLEPHIFPNR SQLCVSAEQS EGHWKVKNCE ERLFYICKKA GHVLSDAESG CQEGWERHGG FCYKID Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 76%.

Receptor for secretory phospholipase A2 (sPLA2). Acts as a receptor for phosholipase sPLA2-IB/PLA2G1B but not sPLA2-IIA/PLA2G2A. Also able to bind to snake PA2-like toxins. Although its precise function remains unclear, binding of sPLA2 to its receptor participates in both positive and negative regulation of sPLA2 functions as well as clearance of sPLA2. Binding of sPLA2-IB/PLA2G1B induces various effects depending on the cell type, such as activation of the mitogen-activated protein kinase (MAPK) cascade to induce cell proliferation, the production of lipid mediators, selective release of arachidonic acid in bone marrow-derived mast cells. In neutrophils, binding of sPLA2-IB/PLA2G1B can activate p38 MAPK to stimulate elastase release and cell adhesion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2R1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pla2r1
Gene Accession No. Q13018
Gene Alias 180 kDa secretory phospholipase A2 receptor; CLEC13C; C-type lectin domain family 13 member C; mannose receptor, C type 2; Mrc2; M-type receptor; phospholipase A2 receptor 1; phospholipase A2 receptor 1, 180kD; phospholipase A2 receptor 1, 180kDa; phospholipase A2, group IB, pancreas, receptor; Pla2g1br; PLA2G1R; PLA2-I receptor; PLA2IR; Pla2r; Pla2-r; PLA2R1; Secretory phospholipase A2 receptor; Soluble PLA2-R; Soluble secretory phospholipase A2 receptor
Gene Symbols Pla2r1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLA2R1. Recombinant protein control fragment (Product #RP-90366).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22925
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.