Learn More
Invitrogen™ PLA2R1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587610
Description
Immunogen sequence: GASWWIGLQE ERANDEFRWR DGTPVIYQNW DTGRERTVNN QSQRCGFISS ITGLWGSEEC SVSMPSICKR KKVWLIEKKK DTPKQHGTCP KGWLYFNYKC LLLNIPKDPS SWKNWTHAQH FCAEEGGTLV AIESEVEQAF ITMNLFGQTT SVWIGLQNDD YETWLNGKPV VYSNWSPFDI INIPSHNTTE VQKHIPLCAL LSSNPNFHFT GKWYFEDCGK EGYGFVCEKM QDTSGHGVNT SDMYPMPNTL EYGNRTYKII N; Positive Samples: 293T, SH-SY5Y, Mouse liver, Mouse heart; Cellular Location: Cell membrane, Single-pass type I membrane protein.
Receptor for secretory phospholipase A2 (sPLA2). Acts as a receptor for phosholipase sPLA2-IB/PLA2G1B but not sPLA2-IIA/PLA2G2A. Also able to bind to snake PA2-like toxins. Although its precise function remains unclear, binding of sPLA2 to its receptor participates in both positive and negative regulation of sPLA2 functions as well as clearance of sPLA2. Binding of sPLA2-IB/PLA2G1B induces various effects depending on the cell type, such as activation of the mitogen-activated protein kinase (MAPK) cascade to induce cell proliferation, the production of lipid mediators, selective release of arachidonic acid in bone marrow-derived mast cells. In neutrophils, binding of sPLA2-IB/PLA2G1B can activate p38 MAPK to stimulate elastase release and cell adhesion.
Specifications
PLA2R1 | |
Polyclonal | |
Unconjugated | |
Pla2r1 | |
180 kDa secretory phospholipase A2 receptor; CLEC13C; C-type lectin domain family 13 member C; mannose receptor, C type 2; Mrc2; M-type receptor; phospholipase A2 receptor 1; phospholipase A2 receptor 1, 180kD; phospholipase A2 receptor 1, 180kDa; phospholipase A2, group IB, pancreas, receptor; Pla2g1br; PLA2G1R; PLA2-I receptor; PLA2IR; Pla2r; Pla2-r; PLA2R1; Secretory phospholipase A2 receptor; Soluble PLA2-R; Soluble secretory phospholipase A2 receptor | |
Rabbit | |
Affinity Chromatography | |
RUO | |
22925 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.37 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
Q13018 | |
Pla2r1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 870-1130 of human PLA2R1 (NP_0313923). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.