Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLAA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554299
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554299 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554299 Supplier Invitrogen™ Supplier No. PA554299
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TGAGRYVPGS ASMGTTMAGV DPFTGNSAYR SAASKTMNIY FPKKEAVTFD QANPTQILGK LKELNGTAPE EKKLTEDDLI LLEKILSLIC Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

Phospholipase A-2-activating protein (PLAA), also named PLAP, can interact with ubiquitin and is involved in the maintenance of ubiquitin levels. PLAA contains multiple consensus domains including WD repeats, PFU domain and PUL domain. Although the function of PLAA remains unclear, ubiquitin binding of PLAA might be the central role that connects ubiquitination and degradation in ERAD. Recent finding revealed that PLAA served as a novel nociceptive mediator after incision, and its expression level is regulated by miR-203.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLAA
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Plaa
Gene Accession No. P27612, P54319, Q9Y263
Gene Alias 2410007N06; AI536418; AU018445; AW208417; D4Ertd618e; DOA1; DOA1 homolog; phospholipase A2 activating protein; phospholipase A2, activating protein; phospholipase A-2-activating protein; phospholipase A2-activating protein; PLA2P; Plaa; PLAP; Ufd3
Gene Symbols Plaa
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLAA. Recombinant protein control fragment (Product #RP-92984).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116645, 18786, 9373
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.