Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLAC2 Mouse anti-Human, Clone: OTI6C11, lyophilized, TrueMAB™
SDP

Catalog No. 501670086
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-0086 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-0086 Supplier OriGene Supplier No. CF810461
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

PLAC2 (placenta-specific protein 2) is a 151 amino acid protein that is encoded by a gene located on human chromosome 19p13. Based on its amino acid sequence, PLAC2 is predicted to be a multi-pass membrane protein. There are no reported studies or functions reported for this protein.

Specifications

Antigen PLAC2
Applications Western Blot
Classification Monoclonal
Clone OTI6C11
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene TINCR
Gene Alias LINC00036, NCRNA00036, PLAC2, onco-lncRNA-16
Gene Symbols TINCR
Host Species Mouse
Immunogen Full length human recombit protein of human PLAC2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257000
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.