Learn More
PLAC2 Mouse anti-Human, Clone: OTI6G4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810460

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
PLAC2 (placenta-specific protein 2) is a 151 amino acid protein that is encoded by a gene located on human chromosome 19p13. Based on its amino acid sequence, PLAC2 is predicted to be a multi-pass membrane protein. There are no reported studies or functions reported for this protein.Specifications
PLAC2 | |
Monoclonal | |
Unconjugated | |
TINCR | |
TINCR | |
Full length human recombit protein of human PLAC2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI6G4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
LINC00036, NCRNA00036, PLAC2, onco-lncRNA-16 | |
Mouse | |
Affinity Chromatography | |
RUO | |
257000 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.