Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLAC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521047
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521047 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521047 Supplier Invitrogen™ Supplier No. PA521047
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human placenta lysate. PA5-21047 can be used with blocking peptide PEP-1161.

PLAC4 (placenta-specific protein 4), was initially discovered as a gene whose mRNA was expressed in placental syncytiotrophoblast and choriocarcinoma cells. While the function of PLAC4 is currently unclear, its gene is known to be located on chromosome 21q22.3. Due to the PLAC4 gene location on chromosome 21 and the release of its mRNA transcripts into maternal bloodstream from the placenta, it has been proposed to be a useful marker noninvasive prenatal chromosome 21 aneuploidy detection, although other reports suggests that PLAC4 mRNA levels are not altered in the maternal circulation of pregnancies with trisomy 21.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLAC4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene PLAC4
Gene Accession No. Q8WY50
Gene Alias C21orf115; PLAC4; placenta specific 4; placenta-specific 4; placenta-specific protein 4; PRED78
Gene Symbols PLAC4
Host Species Rabbit
Immunogen A 17 amino acid peptide near the center of human PLAC4.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 191585
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.