Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLAC8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559189
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559189 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559189 Supplier Invitrogen™ Supplier No. PA559189
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MQAQAPVVVV TQPGVGPGPA PQNSNWQTGM CDCFSDCGVC LCGTFCFPCL GCQVAADMNE CCLCGTSVAM RTLYRTRYGI PGSICDDYMA TLCCPHCTLC QIKRDINRRR AMRTF Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.

PLAC8 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLAC8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLAC8
Gene Accession No. Q9NZF1
Gene Alias BM-004; C15; D5Wsu111e; DGIC; down-regulated in gastrointestinal cancer protein; onzin; PLAC8; placenta specific 8; placenta-specific 8; placenta-specific gene 8 protein; PNAS-144; Protein C15
Gene Symbols PLAC8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLAC8. Recombinant protein control fragment (Product #RP-88688).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51316
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.