Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLAC9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569517
Description
This target displays homology in the following species: Cow: 92%; Human: 100%; Mouse: 92%; Pig: 92%; Rat: 92%.
By microarray assay to search for mouse genes enriched in placenta, Plac9 was identified as a placenta-enriched gene expressed in mouse 16-cell morula cDNA. By database analysis, human PLAC9, which encodes a deduced 97-amino acid protein, was identified. The deduced 108-amino acid mouse protein has a predicted signal peptide, a coiled-coil domain, and shares 73% identity with human PLAC9. Northern blot analysis of mouse placenta and embryo RNA at 12.5 and 18.5 days postcoitum (dpc) detected predominant expression in placenta.
Specifications
PLAC9 | |
Polyclonal | |
Unconjugated | |
PLAC9 | |
hypothetical protein LOC510496; PLAC9; Plac9a; placenta associated 9; placenta specific 9; placenta specific 9a; placenta-specific 9; placenta-specific protein 9 | |
Rabbit | |
Affinity chromatography | |
RUO | |
219348 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5JTB6 | |
PLAC9 | |
synthetic peptide directed towards the middle region of human PLAC9. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction