Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLAC9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560373
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560373 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560373 Supplier Invitrogen™ Supplier No. PA560373
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EPFSPPRGDS AQSTACDRHM AVQRRLDVME EMVEKTVDHL GTEVKGLLGL LEELAWNLPP GPFSPAPDLL GDGF Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 69%.

By microarray assay to search for mouse genes enriched in placenta, Plac9 was identified as a placenta-enriched gene expressed in mouse 16-cell morula cDNA. By database analysis, human PLAC9, which encodes a deduced 97-amino acid protein, was identified. The deduced 108-amino acid mouse protein has a predicted signal peptide, a coiled-coil domain, and shares 73% identity with human PLAC9. Northern blot analysis of mouse placenta and embryo RNA at 12.5 and 18.5 days postcoitum (dpc) detected predominant expression in placenta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLAC9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLAC9
Gene Accession No. Q5JTB6
Gene Alias hypothetical protein LOC510496; PLAC9; Plac9a; placenta associated 9; placenta specific 9; placenta specific 9a; placenta-specific 9; placenta-specific protein 9
Gene Symbols PLAC9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLAC9. Recombinant protein control fragment (Product #RP-99992).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219348
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.