Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Placental Alkaline Phosphatase Monoclonal Antibody (8A9), Invitrogen™

Catalog No. PIMA514064
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514064 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514064 Supplier Thermo Scientific Supplier No. MA514064

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

MA5-14064 targets Placental Alk Phosphatase in immunohostochemistry (Paraffin) applications and shows reactivity with Human samples. Staining of formalin-fixed tissue sections requires treating the tissue sections in boiling 10 mM citrate buffer, pH 6.0, for 10-20 minutes followed by cooling at room temperature for 20 minutes. The MA5-14064 immunogen is purified human Placental Alkaline Phosphatase (PLAP).

Placental Alkaline Phosphatase Monoclonal antibody specifically detects Placental Alkaline Phosphatase in Human samples. It is validated for Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Placental Alkaline Phosphatase
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8A9
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation tissue culture supernatant with 0.09% sodium azide
Gene ALPP
Gene Accession No. P05187
Gene Alias Alkaline phosphatase placental type; Alkaline phosphatase Regan isozyme; alkaline phosphatase, placental; alkaline phosphatase, placental type; alkaline phosphomonoesterase; ALP; Alp1; ALPP; AP-TNAP; DOA1; FLJ11281; FLJ40094; FLJ61142; FLJ93059; Germ-cell alkaline phosphatase; glycerophosphatase; HOPS; MGC161443; MGC167935; nagao Isozyme; P PLAP; PALP; PLA2P; PLAA; placental alkaline phosphatase 1; placental heat-stable alkaline phosphatase; PLAP; PLAP1; PLAP-1; rCG23846-like; Regan isozyme; TNAP; TNSALP
Gene Symbols ALPP
Host Species Mouse
Immunogen Purified human Placental Alkaline Phosphatase (PLAP)
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 250
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.