Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Placental Lactogen/CSH1 Antibody (OTI2G4), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273457B
Description
Placental Lactogen/CSH1 Monoclonal specifically detects Placental Lactogen/CSH1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Placental Lactogen/CSH1 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
CSH1 | |
Full length recombinant protein of human Placental Lactogen/CSH1 (NP_072166) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI2G4 | |
Western Blot, Immunohistochemistry | |
Choriomammotropin, chorionic somatomammotropin hormone, chorionic somatomammotropin hormone 1 (placental lactogen), CS-1, CSAchorionic somatomammotropin A, CSH2, CSMT, FLJ75407, hCS-A, Lactogen, Placental lactogen, PLchoriomammotropin | |
Mouse | |
Affinity Purified | |
Cytokine Research, Signal Transduction | |
1442.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction