Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Placental Lactogen/CSH1 Antibody (SR1402), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Placental Lactogen/CSH1 |
---|---|
Clone | SR1402 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
Placental Lactogen/CSH1 Monoclonal antibody specifically detects Placental Lactogen/CSH1 in Human samples. It is validated for Western BlotSpecifications
Placental Lactogen/CSH1 | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Choriomammotropin, chorionic somatomammotropin hormone, chorionic somatomammotropin hormone 1 (placental lactogen), CS-1, CSAchorionic somatomammotropin A, CSH2, CSMT, FLJ75407, hCS-A, Lactogen, Placental lactogen, PLchoriomammotropin | |
A synthesized peptide derived from human Placental Lactogen/CSH1 (Uniprot #: P01243) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1402 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cytokine Research, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
1442 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title