Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Placental Lactogen/CSH1 Antibody (OTI2G4), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273457UV
Description
Placental Lactogen/CSH1 Monoclonal specifically detects Placental Lactogen/CSH1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Placental Lactogen/CSH1 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CSH1 | |
| Full length recombinant protein of human Placental Lactogen/CSH1 (NP_072166) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI2G4 | |
| Western Blot, Immunohistochemistry | |
| Choriomammotropin, chorionic somatomammotropin hormone, chorionic somatomammotropin hormone 1 (placental lactogen), CS-1, CSAchorionic somatomammotropin A, CSH2, CSMT, FLJ75407, hCS-A, Lactogen, Placental lactogen, PLchoriomammotropin | |
| Mouse | |
| Affinity Purified | |
| Cytokine Research, Signal Transduction | |
| 1442.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction