Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Placental Lactogen (hPL) Rabbit Polyclonal Antibody, Epredia™

Catalog No. RB9067P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB9067P 1 mL Concentrated
RB9067P0 100 μL Concentrated
RB9067R1 1 mL Prediluted
RB9067R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RB9067P Supplier Epredia Supplier No. RB9067P

Provide accurate, reproducible results with the Epredia™ Placental Lactogen (hPL), Rabbit Polyclonal Antibody.

Human placental lactogen (hPL) is known to originate in the syncytiotrophoblasts. Ultrastructural localization indicates that placental lactogen is stored in small morphologically distinct cytoplasmic granules within these cells and has a different secretory mechanism from other hormonal peptides such as human chorionic gonadotropin. The level of expression of placental lactogen through pregnancy increases gradually to term.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not known

Immunogen: Purified human placental lactogen

Molecular Weight: 21kDa

Positive Control: hPL recombinant protein, Placenta

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Specifications

Antigen Placental Lactogen (hPL)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Clone Polyclonal
Concentration 200µg/mL
Conjugate Unconjugated
Host Species Rabbit
Immunogen Purified human placental lactogen
Quantity 1 mL
Regulatory Status IVD
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.