Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Placental lactogen Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119820
This item is not returnable.
View return policy
Description
Positive Control: Human placenta tissue lysates, human placenta tissue. Subcellular Location: Secreted.
The human growth hormone (GH) locus is comprised by two GH (GH1 and GH2) genes and three chorionic somatomammotropin (CSH1, CSH2 and CSH-L) genes. Placental lactogens (CSH1, CSH2) are produced in the mammalian placenta and are similar in structure and function to growth hormones. Together, placental lactogens and growth factors play an essential role to assure successful lactation after pregnancy. Placental lactogens also modify the metabolic state of the mother during pregnancy to supply energy to the fetus. Both CSH1 and CSH2 can be detected by this antibody.
Specifications
| Placental lactogen | |
| Polyclonal | |
| Unconjugated | |
| CSH1 | |
| CSH1, CSH2 | |
| Recombinant protein within human Placental lactogen aa 50-217. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| P0DML2, P0DML3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1442, 1443 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction