Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLAG1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA532188
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532188 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532188 Supplier Thermo Scientific Supplier No. PA532188

Rabbit Polyclonal Antibody

Recommended positive controls: PLAG1-transfected 293T, mouse fetal liver. Predicted reactivity: Mouse (97%), Rat (98%), Zebrafish (80%), Chicken (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Pleomorphic adenoma gene 1 encodes a zinc finger protein with 2 putative nuclear localization signals. PLAG1, which is developmentally regulated, has been shown to be consistently rearranged in pleomorphic adenomas of the salivary glands. PLAG1 is activated by the reciprocal chromosomal translocations involving 8q12 in a subset of salivary gland pleomorphic adenomas. Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLAG1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene PLAG1
Gene Accession No. Q6DJT9, Q9QYE0
Gene Alias C2H2 zinc finger protein PLAG1; COL1A2/PLAG1 fusion; HAS2/PLAG1 fusion; Plag1; PLAG1 zinc finger; pleiomorphic adenoma gene 1; pleiomorphic adenoma gene 1 protein; PSA; SGPA; Zinc finger protein PLAG1; ZNF912
Gene Symbols PLAG1
Host Species Rabbit
Immunogen Recombinant fragment corresponding to a region within amino acids 33 and 202 of Human PLAG1
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5324, 56711
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.