Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLAGL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565970
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565970 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565970 Supplier Invitrogen™ Supplier No. PA565970
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QELMKESLQTG DLLSTFHTIS PSFQLKAAAL PPFPLGASAQ NGLASSLPAE VHSLTLSPPE QAAQPMQPLP ESLASLHPSV SPGSP Highest antigen sequence identity to the following orthologs - mouse 50%, rat 47%.

This gene encodes a C2H2 zinc finger protein with transactivation and DNA-binding activities. It has been shown to have anti-proliferative properties, and thus thought to function as a tumor suppressor. In addition, overexpression of this gene during fetal development is believed to underlie the rare disorder, transient neonatal diabetes mellitus (TNDM). This gene is imprinted, with preferential expression of the paternal allele in many tissues, however, biallelic expression has been noted in peripheral blood leucocytes. A recent study reports that tissue-specific imprinting results from variable utilization of monoallelic and biallelic promoters. Many transcript variants differing in the 5' UTR and encoding two different isoforms, have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLAGL1
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Plagl1
Gene Accession No. Q9UM63
Gene Alias Lost on transformation 1; LOT1; LOT-1; PLAG1 like zinc finger 1; PLAGL1; PLAG-like 1; pleiomorphic adenoma gene-like 1; Pleiomorphic adenoma-like protein 1; Tumor suppressor ZAC; ZAC; Zac1; zinc finger protein PLAGL1; zinc finger protein regulator of apoptosis and cell cycle arrest; zinc finger protein Zac 1
Gene Symbols Plagl1
Host Species Rabbit
Immunogen Recombinant Human PLAGL1. Recombinant protein control fragment (Product #RP-105023).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5325
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.