Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLAGL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599049
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599049 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599049 Supplier Invitrogen™ Supplier No. PA599049
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PLAGL2 Polyclonal Antibody for Western Blot, ELISA

The PLAG (Pleiomorphic adenoma gene) family of zinc finger proteins include PLAG1, ZAC1 and PLAG-like 2 (PLAGL2). PLAG1 is the target gene for the pleiomorphic adenomas of the salivary gland. ZAC1 (PLAGL1) concomitantly controls apoptosis and cell cycle arrest through separate pathways. ZAC1 also acts as a transcriptional cofactor for nuclear receptors. PLAGL2 belongs to the krueppel C2H2-type zinc-finger protein family and contains 6 C2H2-type zinc fingers. It functions as a positive regulator of transcription and localizes to the nucleus. PLAGL2 is ubiquitously expressed with particular abundance in spleen, lung and testis, where it may be involved in cell cycle arrest and apoptosis of tumor cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLAGL2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Plagl2
Gene Accession No. Q9UPG8
Gene Alias AU018672; AW552839; C2H2-type zinc finger protein; KIAA0198; mKIAA0198; PLAG1 like zinc finger 2; PLAGL2; pleiomorphic adenoma gene-like 2; pleiomorphic adenoma-like protein 2; pleomorphic adenoma gene like 2; zinc finger protein PLAGL2; ZNF900
Gene Symbols Plagl2
Host Species Rabbit
Immunogen Recombinant Human Zinc finger protein PLAGL2 protein (324-469AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5326
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.