Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Plakophilin 1 Antibody (10B2) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254526100UG
Description
Plakophilin 1 Monoclonal specifically detects Plakophilin 1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.Specifications
Plakophilin 1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, CyTOF-ready | |
B6PBand 6 protein, MGC138829, plakophilin 1 (ectodermal dysplasia/skin fragility syndrome), plakophilin-1 | |
Mouse | |
75 kDa | |
100 μg | |
Cellular Markers | |
5317 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF | |
10B2 | |
Unconjugated | |
PBS with No Preservative | |
PKP1 | |
N-terminus of human Plakophilin-1 protein (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction