Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Plakophilin-3 Antibody (23E3/4), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197674AF647
Description
Plakophilin-3 Monoclonal specifically detects Plakophilin-3 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Plakophilin-3 | |
Monoclonal | |
Alexa Fluor 647 | |
plakophilin 3, plakophilin-3 | |
Mouse | |
Affinity Purified | |
RUO | |
11187 | |
Store at 4C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
23E3/4 | |
Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
PKP3 | |
Clone 23E3/4 is a mouse monoclonal IgG2b antibody derived by fusion of mouse myeloma cells with spleen cells from a mouse immunized with a synthetic peptide corresponding to amino acid residues 570-589 of human plakophilin-3 coupled to keyhole limpet hemocyanin via an additional cysteine residue at the N-terminus (CFTPQSRRLRELPLAADALTF). | |
0.1 mL | |
Primary | |
Human, Mouse, Canine | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction