Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plakophilin 4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP237691 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237691 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP237691 Supplier Novus Biologicals Supplier No. NBP237691
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Plakophilin 4 Polyclonal specifically detects Plakophilin 4 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Plakophilin 4
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunoprecipitation (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_003619.2
Gene Alias catenin 4, FLJ42243, p0071FLJ31261, plakophilin 4, plakophilin-4
Gene Symbols PKP4
Host Species Rabbit
Immunogen This antibody maps to a region between residue 1 to 50 of human Plakophilin 4 using the numbering given in entry NP_003619.2 (GeneID 8502).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8502
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.