Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plakophilin 4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982386
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2386 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2386 Supplier Bioss Supplier No. BS12711R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Plays a role as a regulator of Rho activity during cytokinesis. May play a role in junctional plaques.

Specifications

Antigen Plakophilin 4
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PKP4
Gene Accession No. Q99569
Gene Alias 5031422I09Rik; 9430019K17Rik; Armadillo-related protein; Armrp; catenin 4; p0071; PKP4; plakophilin 4; Plakophilin-4
Gene Symbols PKP4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Plakophilin 4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8502
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.