Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Plamotamab Chimeric Recombinant Human Monoclonal Antibody
GREENER_CHOICE

Catalog No. PIMA542247
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542247 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542247 Supplier Invitrogen™ Supplier No. MA542247
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.

Plamotamab (XmAb13676) is a tumor-targeted antibody that contains both a CD20 binding domain and a cytotoxic T-cell binding domain (CD3). An XmAb Bispecific Fc domain serves as the scaffold for these two antigen binding domains and confers long circulating half-life, stability and ease of manufacture on plamotamab. CD20 is highly expressed on B-cell tumor cells, including chronic lymphocytic leukemia (CLL) and non-Hodgkin lymphoma (NHL) cells. Engagement of CD3 by plamotamab activates T cells for highly potent and targeted killing of CD20-expressing tumor cells. The structural stability and modularity enabled by the XmAb Bispecific Fc domain allowed the tuning of plamotamab's potency to balance anti-tumor activity with the reduction of immune toxicity that can result from T-cell activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Plamotamab Chimeric
Applications ELISA
Classification Recombinant Monoclonal
Concentration 1.9 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias XmAb-13676
Host Species Human
Immunogen Human CD3E/MS4A1.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.