Learn More
Description
- Rectangular shaped plano glass mirrors
- Designed for educational use
- Include silvered back with protective coating
- Pack of 10
Warranty and Services
1 year limited manufacturer warranty.
Specifications
Specifications
| Dimensions (L x W) | 75 x 50 mm (3 x 2 in.) |
| Quantity | 10 Pk. |
| Material | Plano Glass |
| For Use With (Application) | Light, Optics |
| Product Type | Mirror |
| Shape | Rectangular |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
