Learn More
Kemtec™ Plant Cloning II Kit
SCI_ED
Catalog No. S27530 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
9 to 14
Catalog No. Grade
S27530 9 to 14
1 options
Catalog No. S27530 Supplier Kemtec™ Supplier No. 2506
Add to Cart
Add to Cart
Kemtec™ Plant Cloning II Kit allows users to generate a plant by cloning its tissue in an exciting and challenging experience introducing students to biotechnology and sterile technique.
  • Students will establish a shoot multiplication culture using an African violet model, use sterile technique to develop African violet cloned cultures and induce the development of multiple shoots (clones) from the leaf tissue.
  • Resulting plantlets may be started in soil.
  • The complete process takes approximately three months.
  • No hood or incubator is required.
  • Includes an instruction manual and a flash drive with a presentation including images of plant material at various stages of the experiment.
  • Perishable reply card items ship separately upon request and only in the continental US.
  • Meets National Standards for Grade Levels 9 to 14.
Specifications
Grade 9 to 14
Age 14 to 20
Includes Instruction manual, Instructional flash drive, Petri dishes, Spray bottles, Agar, Sucorse, Paring knives, Plastic pipets, Prepaid reply card for perishable medias and plant preservation solution
Weight (English) 4 lb.
Dimensions (L x W x H) 16 x 12 x 6 in.