Learn More
Novus Biologicals™ Plant Hormone Abscisic Acid ELISA Kit (Colorimetric)
Shop All R&D Systems Products

Specifications
Specifications
| Assay Range | 0.156 - 10 ug/mL |
| Assay Sensitivity | 0.04 ug/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Plant Tissue |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Storage Requirements | Store at 4°C. |
| Target | Plant |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.