Learn More
Description
Specifications
Specifications
| Assay Range | 0.625ng/mL to 10ng/mL |
| Assay Sensitivity | 0.467ng/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Plant Tissue |
| Target Species | Plant |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Research Discipline | Lipid and Metabolism |
| Sample Volume | 50 to 100 μL |
| Storage Requirements | Store at 4°C |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
