Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plasma Cell Monoclonal Antibody (LIV3G11), Invitrogen™

Catalog No. PIMA513532
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA513532 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA513532 Supplier Thermo Scientific Supplier No. MA513532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes an intracytoplasmic antigen that is present in normal as well as neoplastic plasma cells. This antibody is of potential value in identifying myeloma or plasmacytoma in bone marrow or other tissues. It may also help differentiate lymphoplasmacytoid lymphoma from lymphocytic and follicular lymphoma.

Plasma Cell Monoclonal antibody specifically detects Plasma Cell in Human samples. It is validated for Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Plasma Cell
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LIV3G11
Concentration 50 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Host Species Mouse
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.