Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Plasma Kallikrein Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA542752
Description
Peptide sequence: VLTAAHCFDG LPLQDVWRIY SGILNLSDIT KDTPFSQIKE IIIHQNYKVS Sequence homology: Dog: 86%; Horse: 86%; Human: 100%; Pig: 79%; Rat: 79%
Plasma Kallikrein Polyclonal antibody specifically detects Plasma Kallikrein in Human samples. It is validated for Western Blot.
Specifications
Plasma Kallikrein | |
Polyclonal | |
Unconjugated | |
Klkb1 | |
antigen, prostate specific; APS; EC 3.4.21.34; Fletcher factor; Kal3; Kal-3; kallikrein; kallikrein 3, plasma; kallikrein B plasma; kallikrein B, plasma (Fletcher factor) 1; kallikrein B, plasma 1; kallikrein B1; Kalp1; KALP15; kininogenin; Klk3; KLKB1; Pk; PKK; PKKD; plasma kallikrein; Plasma kallikrein heavy chain; Plasma kallikrein light chain; Plasma prekallikrein; PPK; prekallikrein; PSA | |
Rabbit | |
Protein A | |
RUO | |
3818 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P03952 | |
Klkb1 | |
Synthetic peptide directed towards the middle region of human KLKB1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction