Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva PlasmidSelect Xtra Medium
GSA_VA

Catalog No. p-8596010 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-100 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-100 Supplier Cytiva Supplier No. 28402402
Only null left
Add to Cart
Add to Cart

PlasmidSelect Xtra is a thiophilic aromatic adsorption chromatography resin with a selectivity that allows supercoiled covalently closed circular forms of plasmid DNA to be separated from open circular forms.

  • PlasmidSelect Xtra allows for generic purification of supercoiled plasmid DNA
  • Allows for consistent purification results when scaling up from research scale to cGMP production
  • Convenient formats for research, process development and quantitative and qualitative analysis
  • The hydrophilic nature of the base matrix ensures low levels of non-specific binding
  • This resin is a BioProcess medium supported for industrial applications
  • Composed of 6% highly cross-linked agarose

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 200 mL
For Use With (Application) Generic Purification of Supercoiled Plasmid DNA
Stability Stable in all commonly used aqueous buffers
Storage Requirements 4 to 30 deg.C, 20% Ethanol
Product Type PlasmidSelect Xtra Media
Particle Size 24 to 44 μm
Grade BioProcess resin
Max. Pressure <120 cm/h
Matrix Cross-linked Agarose
pH Range 3 to 11
Ligand Type 2-mercaptopyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.