Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Plasminogen Monoclonal Antibody (8E7), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA120081
Description
MA1-20081 detects plasminogen in human samples.
Plasminogen is a plasma protein synthesized mostly in the liver. It consists of a single polypeptide chain and has a molecular weight of 88.5 kDa. The plasma concentration is usually in the range of 70 - 200 mg/L. Plasminogen is activated by tissue-plasminogen activator, urokinase and streptokinase, to form plasmin. Activation results from the cleavage and release of the preactivation peptide. Plasmin consists of a heavy (A-) chain which contains 5 kringles and a light (B-) chain with the active site. Plasmin is a trypsin-like protease whose natural substrate is fibrin, with binding sites for fibrin residing on the kringles. Plasmin is essential for fibrinolysis and decreased fibrinolytic potential due to congenital defects in plasmin can lead to recurring thrombosis, thrombophlebitis and pulmonary embolism.
Specifications
Plasminogen | |
Monoclonal | |
9 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P00747 | |
PLG | |
Purified, full length, native, human plasminogen protein. | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Affinity Purification, ELISA, Immunohistochemistry (Paraffin) | |
8E7 | |
Unconjugated | |
PLG | |
Ab1-346; Activation peptide; AI649309; angiostatin; DKFZp779M0222; Pg; plasmin; plasmin heavy chain A; Plasmin heavy chain A, short form; Plasmin light chain B; plasminogen; PLG; RP1-81D8.1 | |
Mouse | |
IgG fraction | |
RUO | |
5340 | |
4° C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction