Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plasminogen (PLG) Monoclonal Antibody (OTI4F5), TrueMAB™, OriGene
SDP

Catalog No. 502544012
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4012 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4012 Supplier OriGene Supplier No. CF810638
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Plasminogen (PLG) Monoclonal antibody specifically detects Plasminogen (PLG) in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Plasminogen (PLG)
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI4F5
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene MYL1
Gene Accession No. P05976
Gene Alias Ab1-346; Activation peptide; AI649309; angiostatin; DKFZp779M0222; Pg; plasmin; plasmin heavy chain A; Plasmin heavy chain A, short form; Plasmin light chain B; plasminogen; PLG; RP1-81D8.1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 2-150 of human MYL1 produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4632
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.