Learn More
Invitrogen™ Plasminogen Polyclonal Antibody

Description
PA1-43053 detects Plasminogen in human samples. PA1-43053 has successfully been used in Western blot and ELISA applications. PA1-43053 was derived from human plasminogen.
Specifications
Specifications
| Antigen | Plasminogen |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 15.9 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50% water with 50% glycerol and no preservative |
| Gene | PLG |
| Gene Accession No. | P00747 |
| Gene Alias | Ab1-346; Activation peptide; AI649309; angiostatin; DKFZp779M0222; Pg; plasmin; plasmin heavy chain A; Plasmin heavy chain A, short form; Plasmin light chain B; plasminogen; PLG; RP1-81D8.1 |
| Gene Symbols | PLG |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.