Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Plasminogen Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA143063
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA143063 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA143063 Supplier Invitrogen™ Supplier No. PA143063
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

PA1-43063 detects Plasminogen in mouse samples. PA1-43063 has successfully been used in Western blot and ELISA applications. PA1-43063 was derived from mouse plasminogen.

Plasminogen is a plasma protein synthesized mostly in the liver. It consists of a single polypeptide chain and has a molecular weight of 88.5 kDa. The plasma concentration is usually in the range of 70 - 200 mg/L. Plasminogen is activated by tissue-plasminogen activator, urokinase and streptokinase, to form plasmin. Activation results from the cleavage and release of the preactivation peptide. Plasmin consists of a heavy (A-) chain which contains 5 kringles and a light (B-) chain with the active site. Plasmin is a trypsin-like protease whose natural substrate is fibrin, with binding sites for fibrin residing on the kringles. Plasmin is essential for fibrinolysis and decreased fibrinolytic potential due to congenital defects in plasmin can lead to recurring thrombosis, thrombophlebitis and pulmonary embolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Plasminogen
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 6.9 mg/mL
Conjugate Unconjugated
Formulation 50% water with 50% glycerol and no preservative
Gene PLG
Gene Accession No. P20918
Gene Alias Ab1-346; Activation peptide; AI649309; angiostatin; DKFZp779M0222; Pg; plasmin; plasmin heavy chain A; Plasmin heavy chain A, short form; Plasmin light chain B; plasminogen; PLG; RP1-81D8.1
Gene Symbols PLG
Host Species Sheep
Immunogen Murine Plasminogen.
Purification Method SDS-PAGE
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18815
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.