Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Plasmodium falciparum HRP-2 Monoclonal Antibody (H36F)
GREENER_CHOICE

Catalog No. PIMA518243
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518243 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518243 Supplier Invitrogen™ Supplier No. MA518243
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes histidine rich protein II (HRP2) of P. falciparum. Purity is >90%. Recommended pairs for ELISA: MA5-18245 or MA5-18246 (capture) and MA5-18243 (conjugated, detection).

TRUSTED_SUSTAINABILITY

Specifications

Antigen Plasmodium falciparum HRP-2
Applications ELISA
Classification Monoclonal
Clone H36F
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias P. falciparum; P. falciparum HRP2; P. falciparum HRP-2
Host Species Mouse
Immunogen Plasmodium falciparum recombinant protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Protozoa
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.