Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Plasmodium falciparum Monoclonal Antibody
GREENER_CHOICE

Catalog No. MA127094
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA127094 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA127094 Supplier Invitrogen™ Supplier No. MA127094
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Plasmodium falciparum is one of four species of Plasmodium which causes malaria in humans. Despite advances in the treatment of malaria, the disease remains one of the most important infectious diseases affecting humans. Malaria is the cause of approximately 2.5 million deaths per year with 200 million to 500 million new cases each year. The treatment of malaria has become more difficult with the emergence of drug resistant strains of Plasmodium. In particular, the widespread resistance of Plasmodium falciparum to chloroquine, one of the most effective dugs, has spurred the development of alternative drugs for the treatment of malaria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Plasmodium falciparum
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 9.4 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.2
Gene Alias P. falciparum
Host Species Mouse
Immunogen Histidine rich protein II from Plasmodium falciparum.
Purification Method Antigen affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Protozoa
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.